Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TGM7 Monoclonal Antibody (602708)
GREENER_CHOICE

Catalog No. PIMA524272
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524272 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524272 Supplier Invitrogen™ Supplier No. MA524272
Add to Cart
Add to Cart

Rat Monoclonal Antibody

In direct ELISAs, no cross-reactivity with recombinant human TGM2, 3, 4, or recombinant mouse TGM2 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

Transglutaminases (TGM; EC 2.3.2.13) are a family of structurally and functionally related enzymes that stabilize protein assemblies through the formation of gamma-glutamyl-epsilon lysine crosslinks. For additional background information on transglutaminases, see TGM1 (MIM 190195).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGM7
Applications Western Blot
Classification Monoclonal
Clone 602708
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene TGM7
Gene Accession No. Q96PF1
Gene Alias LOC102552199; Protein-glutamine gamma-glutamyltransferase Z; protein-glutamine gamma-glutamyltransferase Z-like; TG(Z); TGase Z; TGase-7; TGM7; TGMZ; TGMZEC 2.3.2.13; TGz; transglutaminase 7; transglutaminase Z; Transglutaminase-7
Gene Symbols TGM7
Host Species Rat
Immunogen Sf 21-derived recombinant human Transglutaminase 7/TGM7 Asp2-Pro710.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116179
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.