Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TGM7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA551263
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA551263 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA551263 Supplier Invitrogen™ Supplier No. PA551263
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total Transglutaminase 7 / TGM7 protein.

Transglutaminases (TGM; EC 2.3.2.13) are a family of structurally and functionally related enzymes that stabilize protein assemblies through the formation of gamma-glutamyl-epsilon lysine crosslinks. For additional background information on transglutaminases, see TGM1 (MIM 190195).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGM7
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene TGM7
Gene Accession No. Q96PF1
Gene Alias LOC102552199; Protein-glutamine gamma-glutamyltransferase Z; protein-glutamine gamma-glutamyltransferase Z-like; TG(Z); TGase Z; TGase-7; TGM7; TGMZ; TGMZEC 2.3.2.13; TGz; transglutaminase 7; transglutaminase Z; Transglutaminase-7
Gene Symbols TGM7
Host Species Rabbit
Immunogen Synthetic peptide of human TGM7.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116179
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.