Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TGN38 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 9 publications
$243.50 - $499.50
Specifications
Antigen | TGN38 |
---|---|
Dilution | Western Blot 10-50 ug/ml, Immunohistochemistry 10-50 ug/ml, Immunocytochemistry/Immunofluorescence 1:50-1:250 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP103495SS
![]() |
Novus Biologicals
NBP103495SS |
0.1 mg |
Each for $243.50
|
|
|||||
NBP103495
![]() |
Novus Biologicals
NBP103495 |
0.5 mg |
Each for $499.50
|
|
|||||
Description
TGN38 Polyclonal specifically detects TGN38 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
TGN38 | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Golgi Apparatus Markers, Trans golgi Network Markers | |
Contains 0.02% benzalkonium chloride. with No Preservative | |
TGN46TGN38 homolog, TGN48MGC14722, TGN5148, 51kD isoforms), trans-golgi network protein 2, Trans-Golgi network protein TGN51 | |
TGOLN2 | |
IgG | |
Ammonium sulfate precipitation | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 10-50 ug/ml, Immunohistochemistry 10-50 ug/ml, Immunocytochemistry/Immunofluorescence 1:50-1:250 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Monkey | |
Q62314 | |
10618 | |
A synthetic peptide from mouse TGN38 conjugated to blue carrier protein was used as the antigen. | |
Primary | |
Reconstitute 0.5 mg size in 0.5 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the 0.1 mg size is provided in reconstituted format. | |
TGN38 (or TGN46, TGN48, TGN51 in human). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title