Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TGN46 Polyclonal Antibody
GREENER_CHOICE

Catalog No. APP1069
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
APP1069 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. APP1069 Supplier Invitrogen™ Supplier No. PA11069
Edge

This product is either new or inactive, and cannot be ordered online.

Rabbit Polyclonal Antibody

PA1-1069 detects TGN46 in human samples. PA1-1069 has been successfully used in Western blot and immunofluorescence procedures. By Western blot, this antibody detects a ~51 kDa protein representing TGN46 in HEK293 and HeLa cell lysates. The PA1-1069 immunogen is a synthetic peptide corresponding to residues V(252) V P E Q P S W K D H S K P I S(267) of human TGN46. This peptide (Cat. # PEP-291) is available for use in neutralization and control experiments.

IASPP is one of the most evolutionarily conserved inhibitors of p53 (TP53; MIM 191170), whereas ASPP1. and ASPP2. are activators of p53.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGN46
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene Tgoln2
Gene Accession No. O43493
Gene Alias AU015504; D6Ertd384e; hTGN48; hTGN51; Tgn38; TGN38 homolog; TGN38A; TGN38B; TGN41; TGN46; TGN48; TGN51; Tgoln1; Tgoln2; Trans-Golgi network glycoprotein 46; Trans-Golgi network glycoprotein 48; Trans-Golgi network glycoprotein 51; Trans-Golgi network integral membrane protein 1; trans-Golgi network integral membrane protein 2; Trans-Golgi network integral membrane protein TGN38; trans-golgi network protein; trans-Golgi network protein (46, 48, 51kD isoforms); trans-golgi network protein 1; trans-golgi network protein 2; trans-Golgi network protein TGN51; Ttgn1; TTGN2
Gene Symbols Tgoln2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues V(252) V P E Q P S W K D H S K P I S(267) of human TGN46.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10618
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.