Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TGN46 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA184496
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA184496 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA184496 Supplier Invitrogen™ Supplier No. PA184496
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

For IHC applications, fixation with methanol or methanol/acetone is recommended. Sheep anti Rat TGN38 antibody recognizes rat TGN38, a 357 amino acid single pass trans membrane glycoprotein found primarily in the trans-golgi network, and acts as an excellent marker for this cellular organelle (Humphrey et al.

IASPP is one of the most evolutionarily conserved inhibitors of p53 (TP53; MIM 191170), whereas ASPP1. and ASPP2. are activators of p53.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGN46
Applications Immunohistochemistry (Frozen), Immunomicroscopy, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation PBS with 1% BSA, 25% glycerol and 0.09% sodium azide
Gene Tgoln2
Gene Accession No. P19814, Q62314
Gene Alias AU015504; D6Ertd384e; hTGN48; hTGN51; Tgn38; TGN38 homolog; TGN38A; TGN38B; TGN41; TGN46; TGN48; TGN51; Tgoln1; Tgoln2; Trans-Golgi network glycoprotein 46; Trans-Golgi network glycoprotein 48; Trans-Golgi network glycoprotein 51; Trans-Golgi network integral membrane protein 1; trans-Golgi network integral membrane protein 2; Trans-Golgi network integral membrane protein TGN38; trans-golgi network protein; trans-Golgi network protein (46, 48, 51kD isoforms); trans-golgi network protein 1; trans-golgi network protein 2; trans-Golgi network protein TGN51; Ttgn1; TTGN2
Gene Symbols Tgoln2
Host Species Sheep
Immunogen Recombinant protein corresponding to extracellular domain of TGN38.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192152, 22135
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.