Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TGN46 Recombinant Rabbit Monoclonal Antibody (9G5X6)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA537930
Description
Positive Samples: HeLa Immunogen sequence: MRFVVALVLL NVAAAGAVPL LATESVKQEE AGVRPSAGNV STHPSLSQRP GGSTKSHPEP QTPKDSPSKS SAEAQTPEDT PNKSGAEAKT QKDSSNKSGA.
IASPP is one of the most evolutionarily conserved inhibitors of p53 (TP53; MIM 191170), whereas ASPP1. and ASPP2. are activators of p53.
Specifications
TGN46 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43493 | |
Tgoln2 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human TGN46/TGOLN2 (O43493). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
9G5X6 | |
Unconjugated | |
Tgoln2 | |
AU015504; D6Ertd384e; hTGN48; hTGN51; Tgn38; TGN38 homolog; TGN38A; TGN38B; TGN41; TGN46; TGN48; TGN51; Tgoln1; Tgoln2; Trans-Golgi network glycoprotein 46; Trans-Golgi network glycoprotein 48; Trans-Golgi network glycoprotein 51; Trans-Golgi network integral membrane protein 1; trans-Golgi network integral membrane protein 2; Trans-Golgi network integral membrane protein TGN38; trans-golgi network protein; trans-Golgi network protein (46, 48, 51kD isoforms); trans-golgi network protein 1; trans-golgi network protein 2; trans-Golgi network protein TGN51; Ttgn1; TTGN2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
10618 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction