Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGOLN2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501567709
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-7709 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-7709 Supplier Bethyl Laboratories, Inc Supplier No. A304434A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.

The TGOLN2 protein may be involved in regulating membrane traffic to and from the trans-Golgi network. There are several isoforms, which are produced by alternative splicing. The upregulation of TGOLN2 has been indicated in association with suicide cases. However, protein analysis has not verified that there is any direct correlation between suicide and TGOLN2 without alcohol influence. It is a highly glycosylated protein.

Specifications

Antigen TGOLN2
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene TGOLN2
Gene Accession No. O43493
Gene Alias TGN51, TGN48, Trans Golgi Network Integral Membrane Protein 2, TTGN2
Gene Symbols TGOLN2
Host Species Rabbit
Immunogen Between 25 and 75
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10618
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.