Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TGR5/GPBAR1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
Antigen | TGR5/GPBAR1 |
---|---|
Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
TGR5/GPBAR1 Polyclonal antibody specifically detects TGR5/GPBAR1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
TGR5/GPBAR1 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
GPCR | |
PBS (pH 7.2), 40% Glycerol | |
151306 | |
IgG | |
Immunogen affinity purified |
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
BG37hBG37, G protein-coupled bile acid receptor 1, GPCR19, GPR131, G-protein coupled bile acid receptor 1, G-protein coupled bile acid receptor BG37, G-protein coupled receptor GPCR19, M-BARhGPCR19, membrane bile acid receptor, Membrane-type receptor for bile acids, MGC40597, TGR5GPCR | |
This antibody was developed against a recombinant protein corresponding to amino acids: LGDQRYTAPWRAAAQRCLQGLWGRASRD | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title