Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TGR5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA527076
Description
Recommended positive controls: A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the G protein-coupled receptor (GPCR) superfamily. This enzyme functions as a cell surface receptor for bile acids. Treatment of cells expressing this GPCR with bile acids induces the production of intracellular cAMP, activation of a MAP kinase signaling pathway, and internalization of the receptor. The receptor is implicated in the suppression of macrophage functions and regulation of energy homeostasis by bile acids. Alternative splicing results in multiple transcript variants encoding the same protein.
Specifications
TGR5 | |
Polyclonal | |
Unconjugated | |
GPBAR1 | |
BG37; C16DUPq36.3; DUP7q36.3; G protein-coupled bile acid receptor 1; G protein-coupled receptor; G protein-coupled receptor TGR5; Gpbar1; GPCR; GPCR HBG37; GPCR19; GPR131; G-protein coupled bile acid receptor 1; G-protein coupled bile acid receptor BG37; G-protein coupled receptor GPCR19; hBG37; Helodermin-preferring VIP receptor; hGPCR19; M-BAR; membrane bile acid receptor; membrane-type receptor for bile acids; PACAP type III receptor; PACAP-R3; PACAP-R-3; pituitary adenylate cyclase-activating polypeptide type III receptor; Tgr5; vasoactive intestinal peptide receptor 2; vasoactive intestinal polypeptide receptor 2; vasopressive intestinal peptide receptor 2; VIP and PACAP receptor 2; VIP receptor subtype 2; Vip2; VIP2R; VIPR2; VIP-R-2; VPAC2; VPAC2 receptor; VPAC2R; VPCAP2R | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
151306 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8TDU6 | |
GPBAR1 | |
Synthetic peptide corresponding to a region within amino acids 264 and 330 of Human TGR5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction