Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TH 1760
GREENER_CHOICESDP

Catalog No. 742250 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-225-0 50 mg
74-221-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 74-225-0 Supplier Tocris Bioscience™ Supplier No. 7422/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective NUDT15 hydrolase inhibitor

TH 1760 is a potent and selective NUDT15 (nudix hydrolase 15) inhibitor (IC50 = 29 nM). Displays selectivity for NUDT15 when assayed at 100 μM against other Nudix enzymes (MTH1, NUDT2, 5, 9, 12, 14, 18, 22) and pyrophosphatases or at 10 μM when assayed with a Eurofins Cerep Safety Screen44 panel. In vitro, TH 1760 treatment sensitizes cancer cell lines (NB4 and HL-60) to thiopurine.

Specifications

Chemical Name or Material 6-[[4-(1 H-Indol-5-ylcarbonyl)-1-piperazinyl]sulfonyl]-2(3 H)-benzoxazolone
CAS 2567914-01-4
Quantity 50 mg
Target Hydrolase Inhibitors
Molecular Formula C20H18N4O5S
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.