Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TH Rabbit anti-Human, Unconjugated, Clone: GOO-20, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB20825
Description
TH Monoclonal antibody specifically detects TH in Human samples. It is validated for ICC, IHC, WB-Ce, IF, Flow Cyt.Specifications
TH | |
Monoclonal | |
Unconjugated | |
TH | |
Rabbit | |
100 μg | |
7054 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Immunocytochemistry, Immunohistochemistry, Western Blot, Immunofluorescence, Flow Cytometry | |
GOO-20 | |
tyrosine hydroxylase | |
TYH | |
A synthetic peptide corresponding to human TH | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction