Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TH1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565631
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565631 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565631 Supplier Invitrogen™ Supplier No. PA565631
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ETVENHLKSLL IKHFDPRKAD SIFTEEGETP AWLEQMIAHT TWRDLFYKLA EAHPDCLMLN FTVKLISDAG YQGEITSVS Highest antigen sequence identity to the following orthologs - mouse 100%, rat 99%.

The NELF complex of proteins interacts with the DSIF protein complex to repress transcriptional elongation by RNA polymerase II. The protein encoded by this gene is an essential part of the NELF complex. Alternative translation initiation site usage results in the formation of two isoforms with different N-termini.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TH1L
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NELFCD
Gene Accession No. Q8IXH7
Gene Alias 2410003I03Rik; C77797; HSPC130; Negative elongation factor C/D; negative elongation factor complex member C/D; negative elongation factor complex member C/D, Th1l; negative elongation factor complex member C/D,Th1l; negative elongation factor D; negative elongation factor D-like; negative elongation factor proteins C and D; NELF-C; NELF-C/D; NELFCD; Nelfd; NELF-D; TH1; TH1 drosophila homolog; TH1 homolog; TH1L; TH1-like; TH1-like homolog; TH1-like protein; trihydrophobin 1; trihydrophobin 1 homolog; zgc:55482
Gene Symbols NELFCD
Host Species Rabbit
Immunogen Recombinant Human TH1L. Recombinant protein control fragment (Product #RP-104909).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51497
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.