Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ THADA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545630
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545630 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545630 Supplier Invitrogen™ Supplier No. PA545630
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SQVLYKLEQS KSKREPENEL TKQHPSVSLQ QYKNFMSSIC NSLFEALFPG Sequence homology: Cow: 79%; Dog: 79%; Guinea Pig: 85%; Horse: 79%; Human: 100%; Rabbit: 79%.

THADA (thyroid adenoma associated), also known as GITA, is a 1,953 amino acid protein that is expressed in testis, thyroid, pancreas, stomach and small intestine. Chromosomal aberrations in the gene encoding THADA are associated with benign thyroid adenomas, suggesting a potential role for THADA in tumorigenesis. The THADA gene maps to human chromosome 2, which houses over 1,400 genes and comprises nearly 8% of the human genome. Harlequin icthyosis, a rare and morbid skin deformity, is associated with mutations in the chromosome 2-localized ABCA12 gene, while the lipid metabolic disorder sitosterolemia is associated with defects in the ABCG5 and ABCG8 genes, which also map to chromosome 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen THADA
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene THADA
Gene Accession No. Q6YHU6
Gene Alias 6530405K19; A930021O22; ARMC13; AU019899; BC052885; death receptor-interacting protein; gene inducing thyroid adenomas protein; GITA; KIAA1767; putative death receptor-interacting protein; RGD1560519; Thada; THADA armadillo repeat containing; THADA, armadillo repeat containing; thyroid adenoma associated; thyroid adenoma-associated protein; Thyroid adenoma-associated protein homolog
Gene Symbols THADA
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human THADA (aa 697-746).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63892
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.