Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
THADA, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA557192
Description
The function of this protein remains unknown.
Immunogen sequence: AHGHLQSATD TWENLVSDAR IKQGLIHQHC QVRIDTLGLL CESNRSTEIV SMEEMQWIQF FITYNLNSQS PGVRQQICSL LKKLFCRIQE SSQHighest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 88%.
Specifications
THADA | |
Polyclonal | |
Unconjugated | |
THADA | |
GITA, death receptor-interacting protein, gene inducing thyroid adenomas protein, thyroid adenoma-associated protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
63892 | |
Store at 4°C short term. Store at −20°C long term. Avoid freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6YHU6 | |
THADA | |
Recombinant protein corresponding to Human THADA | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction