Learn More
Description
PROTAC comprising the cyclin-dependent kinase inhibitor SNS 032 (Cat. No. 4075) conjugated to the cereblon E3 ligase ligand, thalidomide (Cat. No. 0652). Potent, selective and cereblon-dependent degrader of Cdk9 (EC50 = 4 nM). Displays >15-fold selectivity for Cdk9 over other CDKs (EC50 values are 62, 171 and 398 nM for Cdk2, Cdk1 and Cdk7, respectively). Induces complete degradation of Cdlk9 at 250 nM in MOLT4 cells. Inhibits proliferation of leukemia cell lines.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | N-(5-(((5-(tert-Butyl)oxazol-2-yl)methyl)thio)thiazol-2-yl)-1-(14-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)-2-oxo-6,9,12-trioxa-3-azatetradecyl)piperidine-4-carboxamide |
| CAS | 2139287-33-3 |
| Quantity | 5 mg |
| Molecular Formula | C40H52N8O10S2 |
| Formula Weight | 869.02 |
| Purity | 98% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

