Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thalidomide
Click to view available options
Quantity:
100 mg
Description
Teratogenic compound found to inhibit HIV-1 replication and FGF-induced angiogenesis
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Quantity | 100 mg |
| UN Number | UN2811 |
| Merck Index | 14,9255 |
| Solubility Information | Soluble in DMSO at 20mg/ml |
| Formula Weight | 258.23 |
| Physical Form | Powder |
| Chemical Name or Material | Thalidomide |
Safety and Handling
| Hazard Category | H301-H312-H340-H360 |
| Hazard Statement | GHS H Statement H301-H340-H360-H312 Toxic if swallowed. May cause genetic defects. May damage fertility or the unborn child. Harmful in contact with skin. |
| Precautionary Statement | P201-P202-P264b-P270-P281-P301+P310-P302+P352-P308+P313-P312-P330-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
| EINECSNumber | 200-031-1 |
| RTECSNumber | TI4375000 |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction