Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-Thalidomide, 99+%

Catalog No. aaj62266mc
Click to view available options
Quantity:
100 mg

CAS: 50-35-1 | C13H10N2O4 | 258.23 g/mol

Teratogenic compound found to inhibit HIV-1 replication and FGF-induced angiogenesis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-35-1
Molecular Formula C13H10N2O4
Molecular Weight (g/mol) 258.23
MDL Number MFCD00153873
InChI Key UEJJHQNACJXSKW-UHFFFAOYNA-N
Synonym thalidomide, thalomid, +/--thalidomide, sedoval, 2-2,6-dioxopiperidin-3-yl isoindoline-1,3-dione, corronarobetin, psycholiquid, psychotablets, theophilcholine, algosediv
PubChem CID 5426
ChEBI CHEBI:74947
IUPAC Name 2-(2,6-dioxopiperidin-3-yl)-2,3-dihydro-1H-isoindole-1,3-dione
SMILES O=C1N(C2CCC(=O)NC2=O)C(=O)C2=CC=CC=C12

Specifications

Color White
Quantity 100 mg
UN Number UN2811
Merck Index 14,9255
Solubility Information Soluble in DMSO; Insoluble in water or ethanol
Formula Weight 258.23
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material (+/-)-Thalidomide
Hazard Category H301-H312-H340-H360
Hazard Statement GHS H Statement
H301-H340-H360-H312
Toxic if swallowed.
May cause genetic defects.
May damage fertility or the unborn child.
Harmful in contact with skin.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P310-P302+P352-P308+P313-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 200-031-1
RTECSNumber TI4375000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.