Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thallium(I) fluoride, 97%

Catalog No. p-7080347
Click to view available options
Quantity:
25 g
100 g

CAS: 7789-27-7 | FTl | 223.38 g/mol

Thallium(I) fluoride is used as chemical reagents, fine chemicals, pharmaceutical research and development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-27-7
Molecular Formula FTl
Molecular Weight (g/mol) 223.38
MDL Number MFCD00049607
InChI Key CULOEOTWMUCRSJ-UHFFFAOYSA-M
Synonym thallium i fluoride, thallium fluoride, thallium monofluoride, thallium fluoride ic
PubChem CID 62675
SMILES [F-].[Tl+]

Specifications

Boiling Point 655°C
Physical Form Powder
Quantity 25 g
UN Number UN1707
Merck Index 14,9262
Solubility Information Very soluble in water
IUPAC Name λ¹-thallanylium fluoride
Formula Weight 223.37
Percent Purity 97%
Density 8.36 g/mL
Chemical Name or Material Thallium (I) fluoride
Show More Show Less
Hazard Category H300+H330-H373
Hazard Statement GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.
EINECSNumber 232-154-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.