Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thallium(I) fluoride, 99%

Catalog No. ac312690050
Click to view available options
Quantity:
5 g
Packaging:
Plastic bottle

CAS: 7789-27-7 | FTl | 223.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-27-7
Molecular Formula FTl
Molecular Weight (g/mol) 223.38
MDL Number MFCD00049607
InChI Key CULOEOTWMUCRSJ-UHFFFAOYSA-M
Synonym thallium i fluoride, thallium fluoride, thallium monofluoride, thallium fluoride ic
PubChem CID 62675
IUPAC Name fluorothallium
SMILES [F-].[Tl+]

Specifications

Boiling Point 655°C
Melting Point 322°C
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 98.5% min
Linear Formula FTl
Merck Index 15, 9411
Solubility Information Solubility in water: 78.6g/100mL (15°C)
Formula Weight 223.37
Percent Purity 99%
Packaging Plastic bottle
Chemical Name or Material Thallium(I) fluoride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Fatal if inhaled.
Fatal if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 232-154-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.