Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thallium(I) sulfate, 99.9+%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass bottle
Plastic bottle
7446-18-6
O4STl2
504.79
MFCD00011278
YTQVHRVITVLIRD-UHFFFAOYSA-L
thallous sulfate,thallium sulfate,thallium i sulfate,tharattin,zelio,bonide antzix,th-universal,unii-u9f9qir12t,rcra waste number p115,sulfuric acid, dithallium 1+ salt
24833
CHEBI:81836
thallium(1+);sulfate
[O-]S(=O)(=O)[O-].[Tl+].[Tl+]
7446-18-6
O4STl2
504.79
MFCD00011278
YTQVHRVITVLIRD-UHFFFAOYSA-L
thallous sulfate,thallium sulfate,thallium i sulfate,tharattin,zelio,bonide antzix,th-universal,unii-u9f9qir12t,rcra waste number p115,sulfuric acid, dithallium 1+ salt
24833
CHEBI:81836
thallium(1+);sulfate
[O-]S(=O)(=O)[O-].[Tl+].[Tl+]
For ultra trace analysis
Assay Percent Range 80.0 to 84% (Tl)
Linear Formula TI2SO4
Merck Index 15,9418
Solubility Information Solubility in water: 4.87g/100mL (20°C)
Formula Weight 504.79
Physical Form Crystals or Powder
Percent Purity 99.9+%
Grade Trace Metal Basis
Density 6.7700g/mL
Packaging Glass bottle
Solubility Solubility in water: 4.87g/100mL (20°C).
Specific Gravity 6.77
Total Trace Metal Impurities 0.1% max
Color White
Melting Point 632.0°C
Quantity 10g
Chemical Name or Material Thallium(I) sulfate, 99.9+%
Show More Show Less

GHS Signal Word: Danger

EINECSNumber : 231-201-3

RTECSNumber : XG6800000

TSCA : TSCA