Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thallium(I) sulfate, 99.9+%, (trace metal basis)

Catalog No. p-221691
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Plastic bottle

CAS: 7446-18-6 | O4STl2 | 504.79 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-18-6
Molecular Formula O4STl2
Molecular Weight (g/mol) 504.79
MDL Number MFCD00011278
InChI Key YTQVHRVITVLIRD-UHFFFAOYSA-L
Synonym thallous sulfate, thallium sulfate, thallium i sulfate, tharattin, zelio, bonide antzix, th-universal, unii-u9f9qir12t, rcra waste number p115, sulfuric acid, dithallium 1+ salt
PubChem CID 24833
ChEBI CHEBI:81836
IUPAC Name thallium(1+);sulfate
SMILES [O-]S(=O)(=O)[O-].[Tl+].[Tl+]

Specifications

Melting Point 632.0°C
Color White
Physical Form Crystals or Powder
Quantity 10 g
Assay Percent Range 80.0 to 84% (Tl)
Linear Formula TI2SO4
Merck Index 15,9418
Solubility Information Solubility in water: 4.87g/100mL (20°C)
Formula Weight 504.79
Percent Purity ≥99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Specific Gravity 6.77
Density 6.7700g/mL
Chemical Name or Material Thallium(I) sulfate, 99.9+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 231-201-3
RTECSNumber XG6800000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.