Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ THAP10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559562
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559562 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559562 Supplier Invitrogen™ Supplier No. PA559562
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HSEAAPGPVS CTRPRAGKQA AASQITCENE LVQTQPHADN PSNTVTSVPT HCEEGPVHKS TQISLKRPRH RSVGIQAKVK AFGKRLCNAT TQ Highest antigen sequence identity to the following orthologs: Mouse - 27%, Rat - 23%.

THAP10 encodes a member of a family of proteins sharing an N-terminal Thanatos-associated domain. The Thanatos-associated domain contains a zinc finger signature similar to DNA-binding domains. This gene is part of a bidirectional gene pair on the long arm of chromosome 15 that is regulated by estrogen and may play a role in breast cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen THAP10
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene THAP10
Gene Accession No. Q9P2Z0
Gene Alias THAP domain containing 10; THAP domain-containing protein 10; THAP10
Gene Symbols THAP10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human THAP10. Recombinant protein control fragment (Product #RP-98060).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56906
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.