Learn More
THAP10 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 218971AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-3627
Only null left
Add to Cart
Description
Description
THAP10 encodes a member of a family of proteins sharing an N-terminal Thanatos-associated domain. The Thanatos-associated domain contains a zinc finger signature similar to DNA-binding domains. This gene is part of a bidirectional gene pair on the long arm of chromosome 15 that is regulated by estrogen and may play a role in breast cancer.
Specifications
Specifications
THAP10
Polyclonal
Unconjugated
THAP10
THAP domain containing 10, THAP10
Rabbit
Antigen Affinity Chromatography
RUO
56906
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.48 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9P2Z0
THAP10
THAP10 Fusion Protein Ag12541
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.