Learn More
Invitrogen™ THAP12 Monoclonal Antibody (1C10-1A6)

Description
Peptide Sequence: MGQLKFNTSE EHHADMYRSD LPNPDTLSAE LHCWRIKWKH RGKDIELPST IYEALHLPDI KFFPNVYALL KVLCILPVMK VENERYENGR KRLKAYLRNT LTDQRSSNLA LLNINFDIKH DLDLMVDTYI KLYTSKSELP TDNSETVENT.
Specifications
Specifications
| Antigen | THAP12 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 1C10-1A6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | THAP12 |
| Gene Accession No. | O43422 |
| Gene Alias | 2900052B10Rik; 52 kDa repressor of the inhibitor of the protein kinase; 58 kDa interferon-induced protein kinase-interacting protein; DAP4; death-associated protein 4; inhibitor of protein kinase PKR; interferon-inducible, double stranded RNA dependent inhibitor, protein-kinase; P52rIPK; p58IPK-interacting protein; P58IPK-regulatory protein; PRKRIR; protein-kinase, interferon-inducible double stranded RNA dependent inhibitor, repressor of (P58 repressor); Rpkrir; testicular tissue protein Li 133; THAP domain containing 12; THAP domain-containing protein 0; THAP domain-containing protein 12; THAP0; THAP12 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.