Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ THAP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596247
Description
Immunogen sequence: CTHIKSMKLK SRNLLKKNNS CSPAGPSNLK SNISSQQVLL EHSYAFRNPM EAKKRIIKLE KEIASLRRKM KTCLQKERRA TRRWIKATCL VKNLEANSVL PKGTSEHMLP TALSSLPLED FKILEQDQQD KTLLSLNLKQ TKSTFI; Positive Samples: Mouse small intestine, Rat brain.
THAP2, also named as THAP domain-containing protein 2, is a 228 amino acid protein, which contains one THAP-type zinc finger. Members of the THAP (thanatos-associated protein) family of proteins contain a well conserved DNA-binding domain known as the THAP-type zinc finger motif. Proteins containing the THAP-type zinc finger motif are commonly involved in transcriptional regulation, cell-cycle control, apoptosis and chromatin modification.
Specifications
THAP2 | |
Polyclonal | |
Unconjugated | |
THAP2 | |
2900040O07Rik; 9030625G08Rik; AI450385; AI649097; THAP domain containing 2; THAP domain containing, apoptosis associated protein 2; THAP domain-containing protein 2; Thap2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
66816, 688019 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
3.04 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q9D305 | |
THAP2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 83-228 of human THAP2 (NP_1136231). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction