Learn More
Invitrogen™ THAP3 Polyclonal Antibody

Description
Keep as concentrated solution. Positive Control: RD. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | THAP3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.52 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | THAP3 |
| Gene Accession No. | Q8WTV1 |
| Gene Alias | 2010013E08Rik; 2210418H06Rik; RGD1308109; snoRNA MBI-15; THAP domain containing 3; THAP domain containing, apoptosis associated protein 3; THAP domain-containing protein 3; THAP3 |
| Gene Symbols | THAP3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.