Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ THAP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588849
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588849 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588849 Supplier Invitrogen™ Supplier No. PA588849
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPKSCAARQC CNRYSSRRKQ LTFHRFPFSR PELLKEWVLN IGRGNFKPKQ HTVICSEHFR PECFSAFGNR KNLKHNAVPT VFAFQDPTQQ VRENTDPASE RGNASSSQKE KVLPEAGAGE DSPGRNMDTA LEELQLPPNA EGHVKQVSPR RPQATEAVGR PTGPAGLRRT PNKQPSDHSY ALLDLDSLKK KLFLTLKENE KLRKRLQAQR LVMRRMSSRL RACKGHQGLQ ARLGPEQQS; Positive Samples: Mouse kidney, Rat liver.

THAP3 (THAP domain-containing protein 3) is a 239 amino acid protein that contains one THAP-type zinc finger and exists as three alternatively spliced isoforms. The gene that encodes THAP3 contains roughly 10,721 bases and maps to human chromosome 1p36.31. Spanning about 260 million base pairs and comprising nearly 8% of the human genome, chromosome 1 is the largest human chromosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen THAP3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene THAP3
Gene Accession No. Q8BJ25
Gene Alias 2010013E08Rik; 2210418H06Rik; RGD1308109; snoRNA MBI-15; THAP domain containing 3; THAP domain containing, apoptosis associated protein 3; THAP domain-containing protein 3; THAP3
Gene Symbols THAP3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-239 of human THAP3 (NP_0011826821).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362667, 69876
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.