Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ THAP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565790
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565790 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565790 Supplier Invitrogen™ Supplier No. PA565790
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DAGDESATSSI EGGVTDKSGI SMDDFTPPGS GACKFIGSLH SYSFSSKHTR ERPSVPREPI DRKRLKKDVE PSCSGSSLGP DKGLAQSPPS SSLT Highest antigen sequence identity to the following orthologs - mouse 74%, rat 73%.

THAP4 gene ontology annotations related to this gene include DNA binding; DNA-binding transcription factor activity, RNA polymerase II-specific; heme binding; metal ion binding; protein homodimerization activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen THAP4
Applications ChIP Assay, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene THAP4
Gene Accession No. Q8WY91
Gene Alias 2010320B01Rik; CGI-36; PP238; THAP domain containing 4; THAP domain-containing protein 4; Thap4
Gene Symbols THAP4
Host Species Rabbit
Immunogen Recombinant Human THAP4. Recombinant protein control fragment (Product #RP-104967).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51078
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.