Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THAP5 Monoclonal Antibody (OTI2D7), TrueMAB™, OriGene
SDP

Catalog No. 501683574
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-3574 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-3574 Supplier OriGene Supplier No. TA810675
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Has sequence-specific DNA-binding activity and can function as transcriptional repressor (in vitro). May be a regulator of cell cycle: THAP5 overexpression in human cell lines causes cell cycle arrest at G2/M phase.

Specifications

Antigen THAP5
Applications Western Blot
Classification Monoclonal
Clone OTI2D7
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene THAP5
Gene Accession No. Q7Z6K1
Gene Alias THAP domain containing 5
Gene Symbols THAP5
Host Species Mouse
Immunogen Full length human recombit protein of human THAP5 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 168451
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.