Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ THAP5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565661
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565661 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565661 Supplier Invitrogen™ Supplier No. PA565661
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EDNQGKDPSKK KSQKKNLEDE KEVCPKAKSE ESFVLNETKK NIVNTDVPHQ HPELLHSSSL VKPPAPKTGS I Highest antigen sequence identity to the following orthologs - mouse 28%, rat 31%.

Has sequence-specific DNA-binding activity and can function as transcriptional repressor (in vitro). May be a regulator of cell cycle: THAP5 overexpression in human cell lines causes cell cycle arrest at G2/M phase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen THAP5
Applications ChIP Assay, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene THAP5
Gene Accession No. Q7Z6K1
Gene Alias THAP domain containing 5; THAP domain-containing protein 5; THAP5
Gene Symbols THAP5
Host Species Rabbit
Immunogen Recombinant Human THAP5. Recombinant protein control fragment (Product #RP-104923).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 168451
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.