Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
THAP6 Antibody (OTI1E10), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274498AF405
Description
THAP6 Monoclonal specifically detects THAP6 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
THAP6 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
THAP6 | |
Full length recombinant protein of human THAP6 (NP_653322) produced in E.coli. | |
0.1 mL | |
152815.0 | |
Store at 4°C in the dark. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI1E10 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
MGC30052, THAP domain containing 6, THAP domain-containing protein 6 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction