Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
THAP6 Antibody (OTI1F6) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274497
Description
THAP6 Monoclonal specifically detects THAP6 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
THAP6 | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000, Immunohistochemistry 1:150 | |
MGC30052, THAP domain containing 6, THAP domain-containing protein 6 | |
Mouse | |
25.5 kDa | |
100 μg | |
152815 | |
Human, Rat | |
Lyophilized |
Western Blot, Immunohistochemistry | |
OTI1F6 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
THAP6 | |
Full length human recombinant protein of human THAP6 (NP_653322) produced in E.coli. | |
Affinity Purified | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction