Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ THAP9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557946
Description
Immunogen sequence: RTWLSKCQPS PGFNSNIFSF LQRRVENGDQ LYQYCSLLIK SIPLKQQLQW DPSSHSLQGF MDFGLGKLDA DETPLASETV LLMA Highest antigen sequence identity to the following orthologs: Mouse - 27%, Rat - 25%.
THAP9 (THAP domain-containing protein 9) is a 903 amino acid protein that contains one THAP-type zinc finger. The gene that encodes THAP9 contains roughly 19,448 bases and maps to human chromosome 4q21.22. Chromosome 4 represents approximately 6% of the human genome and contains nearly 900 genes. Notably, the Huntingtin gene, which is found to encode an expanded glutamine tract in cases of Huntington's disease, is on chromosome 4. FGFR-3 is also encoded on chromosome 4 and has been associated with thanatophoric dwarfism, achondroplasia, Muenke syndrome and bladder cancer. Chromosome 4 is also tied to Ellis-van Creveld syndrome, methylmalonic acidemia and polycystic kidney disease.
Specifications
THAP9 | |
Polyclonal | |
Unconjugated | |
THAP9 | |
DNA transposase THAP9; hTh9; THAP domain containing 9; THAP domain-containing protein 9; THAP9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79725 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H5L6 | |
THAP9 | |
Recombinant protein corresponding to Human THAP9. Recombinant protein control fragment (Product #RP-102765). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction