Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ THAP9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557947
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557947 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557947 Supplier Invitrogen™ Supplier No. PA557947
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSEALLDLSD HRRNLICYAG YVANKLSALL TCEDCITALY ASDLKASKIG SLLFVKKKNG LHFPSESLCR VINICERVVR THSR Highest antigen sequence identity to the following orthologs: Mouse - 23%, Rat - 26%.

THAP9 (THAP domain-containing protein 9) is a 903 amino acid protein that contains one THAP-type zinc finger. The gene that encodes THAP9 contains roughly 19,448 bases and maps to human chromosome 4q21.22. Chromosome 4 represents approximately 6% of the human genome and contains nearly 900 genes. Notably, the Huntingtin gene, which is found to encode an expanded glutamine tract in cases of Huntington's disease, is on chromosome 4. FGFR-3 is also encoded on chromosome 4 and has been associated with thanatophoric dwarfism, achondroplasia, Muenke syndrome and bladder cancer. Chromosome 4 is also tied to Ellis-van Creveld syndrome, methylmalonic acidemia and polycystic kidney disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen THAP9
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene THAP9
Gene Accession No. Q9H5L6
Gene Alias DNA transposase THAP9; hTh9; THAP domain containing 9; THAP domain-containing protein 9; THAP9
Gene Symbols THAP9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human THAP9. Recombinant protein control fragment (Product #RP-96725).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79725
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.