Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thapsigargin, 95%, PROMO, Thermo Scientific Chemicals

Catalog No. J62866MAPM
Click to view available options
Quantity:
10 mg

Applications

Thapsigargin- induces down regulation of the EGF receptor is independent of protein kinase C activation. It is also used as an inducer of stored Ca2+ release.

Solubility

Soluble in DMSO (20 mg/mL), ethanol (20 mg/mL), and acetone.

Notes

Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67526-95-8
Molecular Formula C34H50O12
Molecular Weight (g/mol) 650.76 g/mol
MDL Number MFCD00083511
InChI Key IXFPJGBNCFXKPI-FSIHEZPISA-N
Synonym thapsigargin, unii-z96bq26rzd, z96bq26rzd, chembl96926, octanoic acid 3s-3alpha, 3abeta, 4alpha, 6beta, 6abeta, 7beta, 8alpha z , 9balpha-6-acetyloxy-2,3,-3a,4,5,6,6a,7,8,9b-decahydro-3,3a-dihydroxy-3,6,9-trimethyl-8-2-methyl-1-oxo-2-butenyl oxy-2-oxo-4-1-oxobutoxy-azuleno 4,5-b furan-7-yl ester, octanoic acid, 6-acetyloxy-2,3,3a,4,5,6,6a,7,8,9b-decahydro-3,3a-dihydroxy-3,6,9-trimethyl-8-2-methyl-1-oxo-2-butenyl oxy-2-oxo-4-1-oxobutoxy azuleno 4,5-b furan-7-yl ester, 3s-3alpha,3abeta,4alpha,6beta,6abeta,7beta,8alpha z ,9balpha, tg1, --thapsigargin, octanoic acid 3s-3alpha, 3abeta, 4alpha, 6beta,6abeta, 7beta, 8alpha z , 9balpha-6-acetyloxy-2,3,-3a,4,5,6,6a,7,8,9b-decahydro-3,3a-dihydroxy-3,6,9-trimethyl-8-2-methyl-1-oxo-2-butenyl oxy-2-oxo-4-1-oxobutoxy-azuleno 4,5-b furan-7-yl ester, octanoic acid 3s-3alpha, 3abeta, 4alpha,6beta, 6abeta, 7beta, 8alpha z , 9balpha-6-acetyloxy-2,3,-3a,4,5,6,6a,7,8,9b-decahydro-3,3a-dihydroxy-3,6,9-trimethyl-8-2-methyl-1-oxo-2-butenyl oxy-2-oxo-4-1-oxobutoxy-azuleno 4,5-b furan-7-yl ester
PubChem CID 126969181
IUPAC Name [(3S,3aR,4S,6S,6aS,7S,8S,9bR)-6-acetyloxy-4-butanoyloxy-3,3a-dihydroxy-3,6,9-trimethyl-8-(2-methylbut-2-enoyloxy)-2-oxo-4,5,6a,7,8,9b-hexahydroazuleno[4,5-b]furan-7-yl] octanoate
SMILES CCCCCCCC(=O)O[C@@H]1[C@@H](OC(=O)C(\C)=C/C)C(C)=C2[C@@H]3OC(=O)[C@@](C)(O)[C@@]3(O)[C@H](C[C@](C)(OC(C)=O)[C@@H]12)OC(=O)CCC

Specifications

Color White
Quantity 10 mg
Sensitivity Light Sensitive
Merck Index 14,9272
Solubility Information Soluble in DMSO, ethanol, and acetone.
Percent Purity 95%
Physical Form Film
Chemical Name or Material Thapsigargin
RTECSNumber RH0325700
TSCA No
Recommended Storage Store at -20°C; Store under Nitrogen

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.