Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA175456
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA175456 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA175456 Supplier Thermo Scientific Supplier No. PA175456
Add to Cart
Add to Cart

THC Polyclonal Antibody

THC Polyclonal Antibody for ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen THC
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias C21H30O2; Delta9-Tetrahydrocannabinol; Delta9-THC; Deltanyne; Dronabinol; Marinol; Tetrahydrocannabinol
Host Species Sheep
Immunogen D8-THC-COOH-BSA.
Purification Method Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.