Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

The Chemist's Companion: A Handbook of Practical Data, Techniques, and References
SCI_ED

Catalog No. s51415
Encompass
Click to view available options
:
The Chemists Companion

Includes

Definitions, values, and usage rules of the newly adopted International System of Units (SI Units)

Provides for the teacher and student an unusual adjunct for use in a broad cross-section of chemistry courses.

  • Covers: Properties of atoms and molecules
  • Spectroscopy
  • Photochemistry
  • Chromatography
  • Kinetics and thermodynamics
  • Various experimental techniques
  • and mathematical and numerical information

Specifications

Format Hardcover Book
Product Type The Chemists Companion: A Handbook of Practical Data, Techniques and References
Includes Definitions, values and usage rules of the newly adopted International System of Units (SI Units)
For Use With (Application) Broad cross-section of chemistry courses and Guide for students and professional chemists
Subject Chemistry
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.