Learn More
Ansell The Lite One™ Surgical Masks
Catalog No. 19166990 Shop All Ansell Products
Change view
Click to view available options
Fastening Type:
Earloops
Ties
Color:
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Fastening Type Color
19-166-990 Earloops -
19-035-442 Ties White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-166-990 Supplier Ansell Supplier No. 62356
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Features a protective three-layer construction for adequate protection when exposure to blood or bodily fluids is not a risk.
  • Non-sterile
  • Latex-free
  • Pleat-style with ties
TRUSTED_SUSTAINABILITY
Specifications
Product Type Medical/Procedural Face Mask
Fastening Type Earloops
Style Pleated
Filter Efficiency 0.99
Pore Size 0.1 μm (PFE), 3 μm (BFE)
Series Lite One™