Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ THEG Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570355
Description
This target displays homology in the following species: Cow: 86%; Dog: 86%; Guinea Pig: 85%; Human: 100%; Mouse: 93%; Pig: 86%; Rat: 93%.
THEG gene ontology annotations related to this gene include spermatogenesis.
Specifications
THEG | |
Polyclonal | |
Unconjugated | |
THEG | |
Cancer/testis antigen 56; CT56; testicular haploid expressed gene; testicular haploid expressed gene product; testicular haploid expressed gene protein; testis-specific; Theg; Theg homolog; theg spermatid protein; THEG1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
51298 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9P2T0 | |
THEG | |
synthetic peptide directed towards the middle region of human THEG. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction