Learn More
Invitrogen™ THEM2 Monoclonal Antibody (169CT7.1.3)

Description
THEM2 Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | THEM2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 169CT7.1.3 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | ACOT13 |
| Gene Accession No. | Q9NPJ3 |
| Gene Alias | 0610006O17Rik; Acot13; Acyl-CoA thioesterase 13; acyl-coenzyme A thioesterase 13; Acyl-coenzyme A thioesterase 13, N-terminally processed; HT012; hypothalamus protein HT012; PNAS-27; Them2; thioesterase superfamily member 2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.