Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ THEM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582705
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582705 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582705 Supplier Invitrogen™ Supplier No. PA582705
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GAPGVSVDMN ITYMSPAKLG EDIVITAHVL KQGKTLAFTS VDLTNKATGK LIAQGRHTKH LGN Highest antigen sequence indentity to the following orthologs: Mouse - 92%, Rat - 90%.

THEM2, also known as ACOT13, belongs to the hotdog-fold superfamily and possesses thioesterase activity, with greater activity observed with longer chain acyl-CoAs such as myristoyl- and palmitoyl-CoA. THEM2 is highly expressed in several tissues such as heart, kidney, liver and brain and has been shown to be co-localized with beta-tubulin on microtubules. THEM2 interacts with StarD, a protein that plays a key role in fatty acid metabolism, and the addition of StarD to THEM2 increased its catalytic activity, suggesting that THEM2 plays a significant role in the metabolism of fatty acids. At least two isoforms of THEM2 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen THEM2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ACOT13
Gene Accession No. Q9NPJ3
Gene Alias 0610006O17Rik; Acot13; Acyl-CoA thioesterase 13; acyl-coenzyme A thioesterase 13; Acyl-coenzyme A thioesterase 13, N-terminally processed; HT012; hypothalamus protein HT012; PNAS-27; Them2; thioesterase superfamily member 2
Gene Symbols ACOT13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ACOT13. Recombinant protein control fragment (Product #RP-91726).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55856
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.