Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ THEM4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520773
Description
A suggested positive control is human liver tissue lysate. PA5-20773 can be used with blocking peptide PEP-0887.
THEM4, also known as CTMP, binds specifically to the carboxy-terminal regulatory domain of PKB/Akt at the plasma membrane and acts as a negative regulator, reversing the phenotype of v-Akt-transformed cells. Hypermethylation of the THEM4 promoter and transcriptional downregulation of the gene has been reported in multiple glioblastomas, suggesting that epigenetic regulation of THEM4 may play a role in the progression of this cancer. Bioinformatic analysis, confirmed by in vitro testing, indicates that THEM4 is a broad-range, high activity acyl-CoA thioesterase. Recent reports have also indicated that TMEM4 is a mitochondrial protein whose overexpression is associated with an increase in mitochondrial membrane depolarization and caspase-3 and PARP cleavage, suggesting that THEM4 is involved in the apoptotic program.
Specifications
THEM4 | |
Polyclonal | |
Unconjugated | |
THEM4 | |
2700077M13Rik; 4921507I02Rik; Acyl-CoA thioesterase THEM4; acyl-coenzyme A thioesterase THEM4; carboxyl-terminal modulator protein; C-terminal modulator protein; CTMP; RGD1304723; THEM4; Thioesterase superfamily member 4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
117145, 361992, 75778 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q3UUI3, Q566R0, Q5T1C6 | |
THEM4 | |
An 18 amino acid peptide near the center of human THEM4. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction