Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ THEM4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110484
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110484 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110484 Supplier Invitrogen™ Supplier No. PA5110484
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSEEVILKDC SVPNPSWNKD LRLLFDQFMK KCEDGSWKRL PSYKRTPTEW IQDFKTHFLD PKLMKEEQMS QAQLFTRSFD DGLGFEYVMF YNDIEKRMVC LFQGGPYLEG PPGF.

THEM4, also known as CTMP, binds specifically to the carboxy-terminal regulatory domain of PKB/Akt at the plasma membrane and acts as a negative regulator, reversing the phenotype of v-Akt-transformed cells. Hypermethylation of the THEM4 promoter and transcriptional downregulation of the gene has been reported in multiple glioblastomas, suggesting that epigenetic regulation of THEM4 may play a role in the progression of this cancer. Bioinformatic analysis, confirmed by in vitro testing, indicates that THEM4 is a broad-range, high activity acyl-CoA thioesterase. Recent reports have also indicated that TMEM4 is a mitochondrial protein whose overexpression is associated with an increase in mitochondrial membrane depolarization and caspase-3 and PARP cleavage, suggesting that THEM4 is involved in the apoptotic program.
TRUSTED_SUSTAINABILITY

Specifications

Antigen THEM4
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene THEM4
Gene Accession No. Q3UUI3, Q5T1C6
Gene Alias 2700077M13Rik; 4921507I02Rik; Acyl-CoA thioesterase THEM4; acyl-coenzyme A thioesterase THEM4; carboxyl-terminal modulator protein; C-terminal modulator protein; CTMP; RGD1304723; THEM4; Thioesterase superfamily member 4
Gene Symbols THEM4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 37-150 of human THEM4 (NP_444283.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117145, 75778
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.