Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ THEM5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551205
Description
The antibody detects endogenous levels of total THEM5 protein.
THEM5 (thioesterase superfamily member 5) is a 247 amino acid protein that belongs to the thioesterase superfamily. The gene that encodes THEM5 contains nearly 8,000 bases and maps to human chromosome 1q21.3. Chromosome 1 is the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes on chromosome 1, and considering the great number of genes there are also a large number of diseases associated with chromosome 1. Notably, the rare aging disease Hutchinson-Gilford progeria is associated with the LMNA gene which encodes lamin A. When defective, the LMNA gene product can build up in the nucleus and cause characteristic nuclear blebs. The mechanism of rapidly enhanced aging is unclear and is a topic of continuing exploration. The MUTYH gene is located on chromosome 1 and is partially responsible for familial adenomatous polyposis. Stickler syndrome, Parkinsons, Gaucher disease and Usher syndrome are also associated with chromosome 1. Has acyl-CoA thioesterase activity towards long-chain (C16 and C18) fatty acyl-CoA substrates, with a preference for linoleyl-CoA and other unsaturated long-chain fatty acid-CoA esters. Plays an important role in mitochondrial fatty acid metabolism, and in remodeling of the mitochondrial lipid cardiolipin. Required for normal mitochondrial function.
Specifications
THEM5 | |
Polyclonal | |
Unconjugated | |
THEM5 | |
ACOT15; Acyl-CoA thioesterase THEM5; acyl-coenzyme A thioesterase 15; Acyl-coenzyme A thioesterase THEM5; THEM5; Thioesterase superfamily member 5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
284486 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8N1Q8 | |
THEM5 | |
Full length fusion protein of human THEM5. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction