Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Themis Antibody (701010), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB6929U
Description
Themis Monoclonal antibody specifically detects Themis in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
Themis | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
387357 | |
Store at 4C in the dark. | |
IgG2a |
Flow Cytometry, CyTOF | |
701010 | |
Intracellular Staining by Flow Cytometry, CyTOF-ready | |
bA325O24.3, bA325O24.4, C6orf190, C6orf207, FLJ40584, GASP, GRB2-associated protein, MGC163388, protein THEMIS, signaling phosphoprotein specific for T cells, thymocyte selection associated, thymocyte selection pathway associated, thymocyte-expressed molecule involved in selection, TSEPA | |
E. coli-derived recombinant human Themis, Ala2-Pro282, Accession # Q8N1K5 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction