Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Themis Antibody (719945), Alexa Fluor™ 405, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB6816V
Description
Themis Monoclonal antibody specifically detects Themis in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
Themis | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
387357 | |
Store at 4C in the dark. | |
IgG2b |
Western Blot, Flow Cytometry, CyTOF | |
719945 | |
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
bA325O24.3, bA325O24.4, C6orf190, C6orf207, FLJ40584, GASP, GRB2-associated protein, MGC163388, protein THEMIS, signaling phosphoprotein specific for T cells, thymocyte selection associated, thymocyte selection pathway associated, thymocyte-expressed molecule involved in selection, TSEPA | |
E. coli-derived recombinant mouse Themis, Lys122-Lys237, Accession # Q8BGW0 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction