Learn More
THEMIS Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305277AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
Themis is a cytoplasmic protein that is essential for positive selection and the development of single-positive thymocytes. Highest levels of expression are found in CD4+CD8+ double-positive thymocytes, with lower expression found in mature T cells. In addition to Grb1, Themis has been found to associate with PLC gamma 1 and LAT and is quickly phosphorylated by SLP-76 following TCR signaling. There are 5 isoforms of mouse Themis that are produced as a result of alternative splicing.Specifications
| THEMIS | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q8N1K5 | |
| THEMIS | |
| Between 591 and 641 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| THEMIS | |
| THEMIS; thymocyte selection associated; C6orf190; C6orf207; FLJ40584; MGC163388; TSEPA; bA325O24.3; bA325O24.4; thymocyte selection pathway associated; thymocyte-expressed molecule involved in selection | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 387357 | |
| 4° C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.