Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THEMIS Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982145
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2145 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2145 Supplier Bioss Supplier No. BS12329R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Themis is a cytoplasmic protein that is essential for positive selection and the development of single-positive thymocytes. Highest levels of expression are found in CD4+CD8+ double-positive thymocytes, with lower expression found in mature T cells. In addition to Grb1, Themis has been found to associate with PLC gamma 1 and LAT and is quickly phosphorylated by SLP-76 following TCR signaling. There are 5 isoforms of mouse Themis that are produced as a result of alternative splicing.

Specifications

Antigen THEMIS
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Themis
Gene Accession No. Q8N1K5
Gene Alias bA325O24.3; bA325O24.4; C6orf190; C6orf207; E430004N04Rik; FLJ40584; Gasp; GRB2-associated protein; Grb2-associating protein; MGC163388; Protein THEMIS; Protein thylex; RGD1560849; signaling phosphoprotein specific for T cells; Spot; Themis; thylex; thymocyte expressed molecule involved in selection; thymocyte selection associated; thymocyte selection pathway associated; Thymocyte-expressed molecule involved in selection; TSEPA
Gene Symbols Themis
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human THEMIS.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387357
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.