Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Themis Recombinant Rabbit Monoclonal Antibody (JE55-73)
GREENER_CHOICE

Catalog No. PIMA542501
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542501 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542501 Supplier Invitrogen™ Supplier No. MA542501
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive Control: Jurkat cell lysates, 293T, rat spleen tissue, human appendix tissue, human spleen tissue, Jurkat. Subcellular Location: Nucleus, Cytoplasm.

Themis is a cytoplasmic protein that is essential for positive selection and the development of single-positive thymocytes. Highest levels of expression are found in CD4+CD8+ double-positive thymocytes, with lower expression found in mature T cells. In addition to Grb1, Themis has been found to associate with PLC gamma 1 and LAT and is quickly phosphorylated by SLP-76 following TCR signaling. There are 5 isoforms of mouse Themis that are produced as a result of alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Themis
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JE55-73
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene Themis
Gene Accession No. Q8N1K5
Gene Alias bA325O24.3; bA325O24.4; C6orf190; C6orf207; E430004N04Rik; FLJ40584; Gasp; GRB2-associated protein; Grb2-associating protein; MGC163388; Protein THEMIS; Protein thylex; RGD1560849; signaling phosphoprotein specific for T cells; SPOT; THEMIS; thylex; thymocyte expressed molecule involved in selection; thymocyte selection associated; thymocyte selection pathway associated; thymocyte-expressed molecule involved in selection; Tsepa
Gene Symbols Themis
Host Species Rabbit
Immunogen Synthetic peptide within C-terminal Human Themis.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387357, 498985
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.