Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thenoyltrifluoroacetone, MP Biomedicals™

Catalog No. ICN15684210 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15684210 10 g
ICN15684225 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15684210 Supplier MP Biomedicals, Inc Supplier No. 0215684210
Add to Cart
Add to Cart

Chemical Identifiers

CAS 326-91-0
Molecular Formula C8H5F3O2S
Molecular Weight (g/mol) 222.18
MDL Number MFCD00005445
InChI Key TXBBUSUXYMIVOS-UHFFFAOYSA-N
Synonym 2-thenoyltrifluoroacetone, thenoyltrifluoroacetone, 1,3-butanedione, 4,4,4-trifluoro-1-2-thienyl, 4,4,4-trifluoro-1-thiophen-2-yl butane-1,3-dione, 4,4,4-trifluoro-1-2-thienyl-1,3-butanedione, 4,4,4-trifluoro-1-2-thienyl butane-1,3-dione, perfluoroacetyl 2-thenoyl methane, alpha-thenoyltrifluoroacetone, 1-thenoyl-3,3,3-trifluoroacetone, .alpha.-thenoyltrifluoroacetone
PubChem CID 5601
IUPAC Name 4,4,4-trifluoro-1-(thiophen-2-yl)butane-1,3-dione
SMILES FC(F)(F)C(=O)CC(=O)C1=CC=CS1

Specifications

Color Yellow
Quantity 10 g
Formula Weight 222.2
Physical Form Solid
Chemical Name or Material Thenoyltrifluoroacetone
EINECSNumber 206-316-7
Recommended Storage Store at 0°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.